Gene Transcripts
- Gene Symbol
- Il17rc
- Entrez Gene ID
- 171095
- Gene Name
- interleukin 17 receptor C
- Gene Aliases
- 1110025H02Rik, Gm19850, IL-17RC, IL17-RC, IL17-RL, Il17rl
- Chromosome Location
- Chr. 6: 113471455 - 113483163 on Build GRCm38
- UniGene ID
- Mm.213397
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK033890.1 |
- |
1 |
3568 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB075652.2 |
- |
1 |
526 |
|
Chr. 6 - 113479455 - 113480121 [-] |
View Full Product Details