Gene Transcripts
- Gene Symbol
- Eed
- Entrez Gene ID
- 13626
- Gene Name
- embryonic ectoderm development
- Gene Aliases
- ENSMUSG00000039373, l(7)5Rn, l7Rn5, lusk
- Chromosome Location
- Chr. 7: 89954654 - 89981419 on Build GRCm38
- UniGene ID
- Mm.380914
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016228.1 |
- |
1 |
373 |
|
AK053947.1 |
- |
|
367 |
|
View Full Product Details