Gene Transcripts
- Gene Symbol
- Foxp1
- Entrez Gene ID
- 108655
- Gene Name
- forkhead box P1
- Gene Aliases
- 3110052D19Rik, 4932443N09Rik, AI461938, AW494214
- Chromosome Location
- Chr. 6: 98925342 - 99523024 on Build GRCm38
- UniGene ID
- Mm.234965
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB202659.1 |
- |
|
105 |
|
Chr. - - [-] |
GenBankmRNA |
AI510098.1 |
- |
1 |
226 |
|
Chr. 6 - 98927511 - 98927987 [-] |
GenBankmRNA |
AA125102.1 |
- |
1 |
44 |
|
Chr. 6 - 98927380 - 98927807 [-] |
GenBankmRNA |
BF658606.1 |
- |
1 |
181 |
|
Chr. 6 - 98927374 - 98927941 [-] |
GenBankmRNA |
BG095300.1 |
- |
1 |
47 |
|
Chr. 6 - 98927449 - 98927810 [-] |
GenBankmRNA |
DT911052.1 |
- |
1 |
57 |
|
Chr. 6 - 98927381 - 98927820 [-] |
View Full Product Details