Gene Transcripts
- Gene Symbol
- Jmjd1c
- Entrez Gene ID
- 108829
- Gene Name
- jumonji domain containing 1C
- Gene Aliases
- 5430433L24Rik, D630035I23Rik, Jmjdic, TRIP8
- Chromosome Location
- Chr. 10: 67096314 - 67256326 on Build GRCm38
- UniGene ID
- Mm.23846
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK087621.1 |
- |
1 |
602 |
|
View Full Product Details