Gene Transcripts
- Gene Symbol
- Epb4.1
- Entrez Gene ID
- 269587
- Gene Name
- erythrocyte protein band 4.1
- Gene Aliases
- 4.1R, AI415518, D4Ertd442e, Elp-1, Elp1, Epb41, mKIAA4056
- Chromosome Location
- Chr. 4: 131923413 - 132075579 on Build GRCm38
- UniGene ID
- Mm.30038
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK148569.1 |
- |
1 |
1867 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ042489.1 |
- |
1 |
108 |
|
Chr. 4 - 131934278 - 131935018 [-] |
View Full Product Details