Gene Transcripts
- Gene Symbol
- Lrrk2
- Entrez Gene ID
- 66725
- Gene Name
- leucine-rich repeat kinase 2
- Gene Aliases
- 4921513O20Rik, 9330188B09Rik, AW561911, D630001M17Rik, Gm927, cI-46
- Chromosome Location
- Chr. 15: 91672821 - 91816124 on Build GRCm38
- UniGene ID
- Mm.37558
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK086917.1 |
- |
2 |
2824 |
|
View Full Product Details