Gene Transcripts
- Gene Symbol
- Ccdc58
- Entrez Gene ID
- 381045
- Gene Name
- coiled-coil domain containing 58
- Gene Aliases
- A930007B11Rik, AI413631
- Chromosome Location
- Chr. 16: 36071660 - 36092118 on Build GRCm38
- UniGene ID
- Mm.310434
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020822.1 |
- |
1 |
382 |
|
AK144564.1 |
- |
1 |
836 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB183663.2 |
- |
1 |
276 |
|
Chr. 16 - 36072186 - 36073114 [+] |
View Full Product Details