Gene Transcripts
- Gene Symbol
- Klhdc8b
- Entrez Gene ID
- 78267
- Gene Name
- kelch domain containing 8B
- Gene Aliases
- 4931406O17Rik
- Chromosome Location
- Chr. 9: 108447636 - 108461581 on Build GRCm38
- UniGene ID
- Mm.283194
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK157991.1 |
- |
1 |
1212 |
|
AK161517.1 |
- |
1 |
361 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CJ047877.1 |
- |
1 |
361 |
|
Chr. 9 - 108459240 - 108460078 [-] |
View Full Product Details