Gene Transcripts
- Gene Symbol
- Klhl3
- Entrez Gene ID
- 100503085
- Gene Name
- kelch-like 3
- Gene Aliases
- 7530408C15Rik, AI430941, EG627648
- Chromosome Location
- Chr. 13: 58004439 - 58123481 on Build GRCm38
- UniGene ID
- Mm.379256
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK033046.1 |
- |
|
2176 |
|
AK082138.1 |
- |
|
55 |
|
View Full Product Details