Gene Transcripts
- Gene Symbol
- Kdelc1
- Entrez Gene ID
- 72050
- Gene Name
- KDEL (Lys-Asp-Glu-Leu) containing 1
- Gene Aliases
- 1810049A15Rik, 5730416C13Rik, EP58, Kdel1
- Chromosome Location
- Chr. 1: 44106546 - 44118857 on Build GRCm38
- UniGene ID
- Mm.281714
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK077468.1 |
- |
1 |
1540 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AV175131.2 |
- |
1 |
494 |
|
Chr. 1 - 44116772 - 44117750 [-] |
View Full Product Details