Gene Transcripts
- Gene Symbol
- Slc30a7
- Entrez Gene ID
- 66500
- Gene Name
- solute carrier family 30 (zinc transporter), member 7
- Gene Aliases
- 1810059J10Rik, 2610034N15Rik, 4833428C12Rik, AI467242, ZnT-7, ZnT7, Zntl2
- Chromosome Location
- Chr. 3: 115938973 - 116007414 on Build GRCm38
- UniGene ID
- Mm.34550
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK076492.1 |
- |
1 |
1063 |
|
AV250971.2 |
- |
1 |
371 |
|
View Full Product Details