Gene Transcripts
- Gene Symbol
- Apopt1
- Entrez Gene ID
- 68020
- Gene Name
- apoptogenic, mitochondrial 1
- Gene Aliases
- 1700081D05Rik, 2810002N01Rik, Apop-1, Apop1
- Chromosome Location
- Chr. 12: 111713269 - 111755055 on Build GRCm38
- UniGene ID
- Mm.425762
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK047131.1 |
- |
1 |
2403 |
|
View Full Product Details