Gene Transcripts
- Gene Symbol
- Gpatch8
- Entrez Gene ID
- 237943
- Gene Name
- G patch domain containing 8
- Gene Aliases
- 5430405G24Rik, AU018890, ENSMUSG00000075516, Fbm1, Gpatc8, mKIAA0553
- Chromosome Location
- Chr. 11: 102475911 - 102556158 on Build GRCm38
- UniGene ID
- Mm.260282
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK084888.1 |
- |
1 |
774 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB481873.2 |
- |
1 |
550 |
|
Chr. 11 - 102555284 - 102555893 [-] |
View Full Product Details