Gene Transcripts
- Gene Symbol
- Klhl24
- Entrez Gene ID
- 75785
- Gene Name
- kelch-like 24
- Gene Aliases
- 1110046J11Rik, 4930429H24Rik, 6530402O20Rik, C85082
- Chromosome Location
- Chr. 16: 20097457 - 20129226 on Build GRCm38
- UniGene ID
- Mm.392450
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK004187.1 |
- |
1 |
175 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY704431.1 |
- |
1 |
175 |
|
Chr. 16 - 20128106 - 20129055 [+] |
View Full Product Details