Gene Transcripts
- Gene Symbol
- March3
- Entrez Gene ID
- 320253
- Gene Name
- membrane-associated ring finger (C3HC4) 3
- Gene Aliases
- 6330411I15Rik, A530081L18Rik, AI843639, MARCH-III
- Chromosome Location
- Chr. 18: 56761716 - 56925652 on Build GRCm38
- UniGene ID
- Mm.312623
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK018160.1 |
- |
1 |
1826 |
|
AK029887.1 |
- |
1 |
1815 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DT908940.1 |
- |
1 |
487 |
|
Chr. 18 - 56923475 - 56924220 [-] |
View Full Product Details