Gene Transcripts

Gene Symbol
Foxo3
Entrez Gene ID
56484
Gene Name
forkhead box O3
Gene Aliases
1110048B16Rik, 2010203A17Rik, C76856, FKHRL1, Fkhr2, Foxo3a
Chromosome Location
Chr. 10: 42181845 - 42276746 on Build GRCm38
UniGene ID
Mm.338613
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK004198.1 - 1 413
AK047922.1 - 1 1234
AK079567.1 - 637
AK159465.1 - 1 3689
BC050975.1 - 381
BC100372.1 - 1 1970

View Full Product Details