Gene Transcripts
- Gene Symbol
- Slamf9
- Entrez Gene ID
- 98365
- Gene Name
- SLAM family member 9
- Gene Aliases
- 2310026I04Rik, AI462096, CD2F-10, CD84-H1, Cd2f10, SF2001
- Chromosome Location
- Chr. 1: 172458348 - 172478421 on Build GRCm38
- UniGene ID
- Mm.150050
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK076693.1 |
- |
|
76 |
|
View Full Product Details