Gene Transcripts
- Gene Symbol
- Stag1
- Entrez Gene ID
- 20842
- Gene Name
- stromal antigen 1
- Gene Aliases
- AU045003, SA-1, Scc3
- Chromosome Location
- Chr. 9: 100597709 - 100958544 on Build GRCm38
- UniGene ID
- Mm.42135
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK034136.1 |
- |
|
2962 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB079646.2 |
- |
1 |
20 |
|
Chr. 9 - 100958710 - 100959372 [+] |
View Full Product Details