Gene Transcripts
- Gene Symbol
- Magef1
- Entrez Gene ID
- 76222
- Gene Name
- melanoma antigen family F, 1
- Gene Aliases
- 6430590I03Rik
- Chromosome Location
- Chr. 16: 21331899 - 21333677 on Build GRCm38
- UniGene ID
- Mm.86031
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK018285.1 |
- |
1 |
929 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB469018.2 |
- |
1 |
142 |
|
Chr. 16 - 21331907 - 21332575 [-] |
View Full Product Details