Gene Transcripts
- Gene Symbol
- Dtymk
- Entrez Gene ID
- 21915
- Gene Name
- deoxythymidylate kinase
- Gene Aliases
- AU044245, CDC8, D10Ertd203e, TMPK, TYPK, Tmk, mtmk
- Chromosome Location
- Chr. 1: 93792576 - 93801934 on Build GRCm38
- UniGene ID
- Mm.250332
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK080078.1 |
- |
2 |
264 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB637195.1 |
- |
2 |
260 |
|
Chr. 1 - 93798440 - 93799349 [+] |
View Full Product Details