Gene Transcripts
- Gene Symbol
- Itprip
- Entrez Gene ID
- 414801
- Gene Name
- inositol 1,4,5-triphosphate receptor interacting protein
- Gene Aliases
- 4833424O12Rik, DANGER, mKIAA1754
- Chromosome Location
- Chr. 19: 47894593 - 47919519 on Build GRCm38
- UniGene ID
- Mm.29457
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK014762.1 |
- |
1 |
1322 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CK626294.1 |
- |
1 |
418 |
|
Chr. 19 - 47917793 - 47918298 [-] |
View Full Product Details