Gene Transcripts
- Gene Symbol
- Tac1
- Entrez Gene ID
- 21333
- Gene Name
- tachykinin 1
- Gene Aliases
- 4930528L02Rik, NK-1, NK1, Nkna, PPT-A, PPTA, SP
- Chromosome Location
- Chr. 6: 7554776 - 7562978 on Build GRCm38
- UniGene ID
- Mm.1440
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015286.1 |
- |
|
16 |
|
AK019709.1 |
- |
|
16 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY715489.1 |
- |
1 |
13 |
|
Chr. 6 - 7562370 - 7564254 [+] |
View Full Product Details