Gene Transcripts
- Gene Symbol
- Slco2a1
- Entrez Gene ID
- 24059
- Gene Name
- solute carrier organic anion transporter family, member 2a1
- Gene Aliases
- 2310021C19Rik, Pgt, Slc21a2, mPgt
- Chromosome Location
- Chr. 9: 103007668 - 103087849 on Build GRCm38
- UniGene ID
- Mm.207106
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK165716.1 |
- |
|
1980 |
|
View Full Product Details