Gene Transcripts
- Gene Symbol
- Exoc3
- Entrez Gene ID
- 211446
- Gene Name
- exocyst complex component 3
- Gene Aliases
- 2810050O03Rik, E430013E20Rik, Sec6, Sec6l1
- Chromosome Location
- Chr. 13: 74169796 - 74208729 on Build GRCm38
- UniGene ID
- Mm.261859
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK012940.1 |
- |
1 |
365 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ560539.1 |
- |
1 |
350 |
|
Chr. 13 - 74208131 - 74208654 [-] |
GenBankmRNA |
BF469786.1 |
- |
1 |
294 |
|
Chr. 13 - 74208287 - 74208598 [-] |
GenBankmRNA |
BY712025.1 |
- |
1 |
365 |
|
Chr. 13 - 74208072 - 74208669 [-] |
GenBankmRNA |
CK341727.1 |
- |
1 |
350 |
|
Chr. 13 - 74208263 - 74208654 [-] |
View Full Product Details