Gene Transcripts
- Gene Symbol
- Gpr55
- Entrez Gene ID
- 227326
- Gene Name
- G protein-coupled receptor 55
- Gene Aliases
- Gm218, Lpir1
- Chromosome Location
- Chr. 1: 85933206 - 85991567 on Build GRCm38
- UniGene ID
- Mm.244158
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK032227.1 |
- |
1 |
2394 |
|
View Full Product Details