Gene Transcripts
- Gene Symbol
- Tbc1d23
- Entrez Gene ID
- 67581
- Gene Name
- TBC1 domain family, member 23
- Gene Aliases
- 4930451A13Rik, AU015720, AU043671, AU043778, D030022P07Rik, W51689
- Chromosome Location
- Chr. 16: 57168858 - 57231532 on Build GRCm38
- UniGene ID
- Mm.257819
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK028852.1 |
- |
1 |
2440 |
|
AK045169.1 |
- |
1 |
149 |
|
AK050825.1 |
- |
1 |
286 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CJ176137.1 |
- |
1 |
106 |
|
Chr. 16 - 57179771 - 57180219 [-] |
GenBankmRNA |
BB440692.2 |
- |
1 |
326 |
|
Chr. 16 - 57179802 - 57180399 [-] |
GenBankmRNA |
CJ171270.1 |
- |
1 |
106 |
|
Chr. 16 - 57179813 - 57180219 [-] |
GenBankmRNA |
BB642953.1 |
- |
1 |
149 |
|
Chr. 16 - 57179712 - 57180261 [-] |
GenBankmRNA |
BB654616.1 |
- |
1 |
286 |
|
Chr. 16 - 57179783 - 57180399 [-] |
View Full Product Details