Gene Transcripts
- Gene Symbol
- Unc13c
- Entrez Gene ID
- 208898
- Gene Name
- unc-13 homolog C (C. elegans)
- Gene Aliases
- 1500037O19Rik, AU019458, D9Ertd414e, Munc13-3, Unc13h3
- Chromosome Location
- Chr. 9: 73479422 - 74008153 on Build GRCm38
- UniGene ID
- Mm.41035
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK005367.1 |
- |
2 |
867 |
|
View Full Product Details