Gene Transcripts
- Gene Symbol
- Vti1a
- Entrez Gene ID
- 53611
- Gene Name
- vesicle transport through interaction with t-SNAREs 1A
- Gene Aliases
- 1110014F16Rik, 1110018K19Rik, 4921537J05Rik, MVti1, MVti1a, Vti1, Vti1-rp2
- Chromosome Location
- Chr. 19: 55316057 - 55627461 on Build GRCm38
- UniGene ID
- Mm.258637
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK003692.1 |
- |
1 |
443 |
|
View Full Product Details