Gene Transcripts
- Gene Symbol
- Sgpp2
- Entrez Gene ID
- 433323
- Gene Name
- sphingosine-1-phosphate phosphotase 2
- Gene Aliases
- SPPase2, Spp2
- Chromosome Location
- Chr. 1: 78309501 - 78420289 on Build GRCm38
- UniGene ID
- Mm.276248
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK087798.1 |
- |
2 |
487 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB666583.1 |
- |
2 |
487 |
|
Chr. 1 - 78327667 - 78340106 [-] |
View Full Product Details