Gene Transcripts
- Gene Symbol
- MICAL3
- Entrez Gene ID
- 57553
- Gene Name
- microtubule associated monooxygenase, calponin and LIM domain containing 3
- Gene Aliases
- MICAL-3
- Chromosome Location
- Chr. 22: 17787649 - 18024559 on Build GRCh38
- UniGene ID
- Hs.528024
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126925.1 |
- |
1 |
186 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000476405.1 |
- |
1 |
186 |
|
Chr. 22 - 17787652 - 17811497 [-] |
View Full Product Details