Gene Transcripts
- Gene Symbol
- AGAP11
- Entrez Gene ID
- 119385
- Gene Name
- ArfGAP with GTPase domain, ankyrin repeat and PH domain 11
- Gene Aliases
- Chromosome Location
- Chr. 10: 86970741 - 87010203 on Build GRCh38
- UniGene ID
- Hs.511787
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB075855.1 |
- |
4 |
2399 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000444431.1 |
- |
4 |
2400 |
|
Chr. 10 - 86992406 - 87010203 [+] |
View Full Product Details