Gene Transcripts
- Gene Symbol
- C15orf54
- Entrez Gene ID
- 400360
- Gene Name
- chromosome 15 open reading frame 54
- Gene Aliases
- Chromosome Location
- Chr. 15: 39250669 - 39254845 on Build GRCh38
- UniGene ID
- Hs.376109
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000561223.1 |
- |
2 |
866 |
|
Chr. 15 - 39250766 - 39252913 [+] |
GENCODE |
ENST00000625107.1 |
- |
|
1059 |
|
Chr. - - [-] |
View Full Product Details