Gene Transcripts
- Gene Symbol
- LINC00471
- Entrez Gene ID
- 151477
- Gene Name
- long intergenic non-protein coding RNA 471
- Gene Aliases
- C2orf52
- Chromosome Location
- Chr. 2: 231508426 - 231514339 on Build GRCh38
- UniGene ID
- Hs.375211
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024079.1 |
|
3 |
1129 |
|
GenBank mRNA |
BC033054.2 |
- |
3 |
1130 |
|
View Full Product Details