Gene Transcripts

Gene Symbol
TSPEAR-AS2
Entrez Gene ID
114043
Gene Name
TSPEAR antisense RNA 2
Gene Aliases
C21orf90
Chromosome Location
Chr. 21: 44517215 - 44518977 on Build GRCh38
UniGene ID
Hs.592163
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC016690.2 - 2 1260
BC064517.1 - 2 1260


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000354333.5 - 2 1260 Chr. 21 - 44517617 - 44520827 [+]

View Full Product Details