Gene Transcripts
- Gene Symbol
- LINC01547
- Entrez Gene ID
- 84536
- Gene Name
- long intergenic non-protein coding RNA 1547
- Gene Aliases
- C21orf67, C21orf69, PRED54
- Chromosome Location
- Chr. 21: 44933284 - 44939913 on Build GRCh38
- UniGene ID
- Hs.534504
- Species
- Homo sapiens
- Gene Symbol
- ENSG00000183250
- Entrez Gene ID
- ENSG00000183250
- Gene Name
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
View Full Product Details