Gene Transcripts

Gene Symbol
C15orf32
Entrez Gene ID
145858
Gene Name
chromosome 15 open reading frame 32
Gene Aliases
Chromosome Location
Chr. 15: 92465762 - 92501117 on Build GRCh38
UniGene ID
Hs.367879
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301106.1 NP_001288035.1 3 1434
NM_153040.2 NP_694585.1 3 1396
GenBank mRNA AK057393.1 - 3 1397
BC109252.2 - 3 1190 40023840


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000556865.1 - 3 1190 Chr. 15 - 92471921 - 92500842 [+]
GENCODE ENST00000624458.1 - 1419 Chr. - - [-]

View Full Product Details