Gene Transcripts
- Gene Symbol
- NSAP11
- Entrez Gene ID
- 100131275
- Gene Name
- nervous system abundant protein 11
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.623961
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY176665.1 |
- |
1 |
131 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000524252.1 |
- |
1 |
131 |
|
Chr. 8 - 18720905 - 18734405 [+] |
View Full Product Details