Gene Transcripts
- Gene Symbol
- C16orf47
- Entrez Gene ID
- 388289
- Gene Name
- chromosome 16 open reading frame 47
- Gene Aliases
- Chromosome Location
- Chr. 16: 73125865 - 73812449 on Build GRCh38
- UniGene ID
- Hs.371893
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207385.1 |
NP_997268.1
|
1, 2 |
146 |
|
GenBank mRNA |
AK129695.1 |
- |
1, 2 |
146 |
|
View Full Product Details