Gene Transcripts
- Gene Symbol
- C9orf41-AS1
- Entrez Gene ID
- 101927380
- Gene Name
- C9orf41 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 9: 74952965 - 74997382 on Build GRCh38
- UniGene ID
- Hs.257556
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_121183.1 |
|
5 |
527 |
|
GenBank mRNA |
BC043649.2 |
- |
5 |
523 |
|
BI552900.1 |
- |
5 |
528 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000455609.1 |
- |
5 |
524 |
|
Chr. 9 - 74952968 - 74996293 [+] |
View Full Product Details