Gene Transcripts

Gene Symbol
C6orf99
Entrez Gene ID
100130967
Gene Name
chromosome 6 open reading frame 99
Gene Aliases
yR211F11.1
Chromosome Location
Chr. 6: 158869865 - 158918115 on Build GRCh38
UniGene ID
Hs.32804
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195032.1 NP_001181961.1 3 593
NM_001302839.1 NP_001289768.1 3 927
NM_001302840.1 NP_001289769.1 4 688
NM_001302841.1 NP_001289770.1 3 587
XM_005266788.4 XP_005266845.1 3 934
XM_017010142.1 XP_016865631.1 3 524
XM_017010143.1 XP_016865632.1 3 601
XM_017010144.1 XP_016865633.1 3 596
XM_017010145.1 XP_016865634.1 2 499
XM_017010146.1 XP_016865635.1 2 494
XM_017010147.1 XP_016865636.1 3 584
GenBank mRNA BF688381.1 - 3 593


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000608817.2 - 4 492 Chr. 6 - 158870191 - 158910352 [+]

View Full Product Details