Gene Transcripts

Gene Symbol
PSORS1C3
Entrez Gene ID
100130889
Gene Name
psoriasis susceptibility 1 candidate 3 (non-protein coding)
Gene Aliases
NCRNA00196
Chromosome Location
Chr. 6: 31173735 - 31177899 on Build GRCh38
UniGene ID
Hs.670091
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026816.1 3 569
GenBank mRNA AB932952.1 - 2 536
AB932953.1 - 3 763
AB932954.1 - 3 567
AB933312.1 - 4 834
AB933313.1 - 4 600
AB933320.1 - 6 1052
AB933321.1 - 4 748
AY484516.1 - 3 569
LC027944.1 - 3 803
LC027945.1 - 4 861
LC027946.1 - 3 828
LC050979.1 - 4 659
LC050980.1 - 3 639
LC050981.1 - 5 1045
LC050982.1 - 5 1126
LC050983.1 - 5 892
LC050984.1 - 3 600
LC050985.1 - 4 721
LC050986.1 - 6 1078
LC050987.1 - 7 1345
LC050988.1 - 2 567
LC050989.1 - 4 859
LC050990.1 - 6 931
LC050991.1 - 4 1095


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000412143.1 - 3 569 Chr. 6 - 31173735 - 31177899 [-]

View Full Product Details