Gene Transcripts

Gene Symbol
ENTPD3-AS1
Entrez Gene ID
285266
Gene Name
ENTPD3 antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 3: 40390951 - 40453308 on Build GRCh38
UniGene ID
Hs.619663
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040100.1 3 328
GenBank mRNA BC047546.1 - 3 325
BG527749.1 - 3 201


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000425156.5 - 3 325 Chr. 3 - 40390951 - 40453305 [-]
GENCODE ENST00000452768.5 - 3 200 Chr. 3 - 40391850 - 40453180 [-]

View Full Product Details