Gene Transcripts
- Gene Symbol
- STK24-AS1
- Entrez Gene ID
- 100874069
- Gene Name
- STK24 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 13: 98577585 - 98578842 on Build GRCh38
- UniGene ID
- Hs.594160
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF339799.1 |
- |
2 |
531 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000434547.1 |
- |
2 |
531 |
|
Chr. 13 - 98577244 - 98578830 [+] |
View Full Product Details