Gene Transcripts
- Gene Symbol
- TNK2-AS1
- Entrez Gene ID
- 100128262
- Gene Name
- TNK2 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.605240
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK127609.1 |
- |
1 |
193 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000458180.1 |
- |
1 |
193 |
|
Chr. 3 - 195908076 - 195911257 [+] |
View Full Product Details