Gene Transcripts
- Gene Symbol
- OPA1-AS1
- Entrez Gene ID
- 100873941
- Gene Name
- OPA1 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 193618599 - 193627332 on Build GRCh38
- UniGene ID
- Hs.679364
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046634.1 |
|
3 |
293 |
|
GenBank mRNA |
BG203156.1 |
- |
1 |
153 |
|
BG204670.1 |
- |
3 |
293 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000444085.1 |
- |
2 |
212 |
|
Chr. 3 - 193618609 - 193627337 [-] |
View Full Product Details