Gene Transcripts
- Gene Symbol
- RSU1
- Entrez Gene ID
- 6251
- Gene Name
- Ras suppressor protein 1
- Gene Aliases
- RSP-1
- Chromosome Location
- Chr. 10: 16590616 - 16817537 on Build GRCh38
- UniGene ID
- Hs.524161
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092036.1 |
- |
1 |
1223 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000421480.2 |
- |
3 |
436 |
|
Chr. 10 - 16721352 - 16748377 [-] |
View Full Product Details