Gene Transcripts
- Gene Symbol
- KDM4C
- Entrez Gene ID
- 23081
- Gene Name
- lysine demethylase 4C
- Gene Aliases
- GASC1, JHDM3C, JMJD2C, TDRD14C
- Chromosome Location
- Chr. 9: 6720863 - 7175648 on Build GRCh38
- UniGene ID
- Hs.709425
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098534.1 |
- |
2 |
267 |
|
BC042976.2 |
- |
2 |
320 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000615168.4 |
- |
|
320 |
|
Chr. - - [-] |
GENCODE |
ENST00000452643.5 |
- |
2 |
267 |
|
Chr. 9 - 6716548 - 6723630 [+] |
View Full Product Details