Gene Transcripts

Gene Symbol
LINC00685
Entrez Gene ID
283981
Gene Name
long intergenic non-protein coding RNA 685
Gene Aliases
CXYorf10, NCRNA00107, PPP2R3B-AS1
Chromosome Location
Chr. Y: 319145 - 321332 on Build GRCh38
UniGene ID
Hs.575741
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027231.1 2 1210
NR_027232.1 1 1866
GenBank mRNA AK023454.1 - 1 1825
BC041934.1 - 2 1213

View Full Product Details