Gene Transcripts
- Gene Symbol
- DNMBP-AS1
- Entrez Gene ID
- 100188954
- Gene Name
- DNMBP antisense RNA 1
- Gene Aliases
- NCRNA00093, bA287G8.2
- Chromosome Location
- Chr. 10: 99927209 - 99958998 on Build GRCh38
- UniGene ID
- Hs.656104
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024130.2 |
|
5 |
1949 |
|
GenBank mRNA |
AK098111.1 |
- |
5 |
1951 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000434409.1 |
- |
5 |
1949 |
|
Chr. 10 - 99927209 - 99958381 [+] |
View Full Product Details