Gene Transcripts
- Gene Symbol
- MFI2-AS1
- Entrez Gene ID
- 100507057
- Gene Name
- MFI2 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 197002906 - 197004744 on Build GRCh38
- UniGene ID
- Hs.666451
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038285.1 |
|
4 |
877 |
|
GenBank mRNA |
BC032042.1 |
- |
4 |
877 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000424769.1 |
- |
2 |
290 |
|
Chr. 3 - 196999460 - 197004736 [+] |
GENCODE |
ENST00000415244.1 |
- |
4 |
877 |
|
Chr. 3 - 197002906 - 197004744 [+] |
GENCODE |
ENST00000414354.1 |
- |
2 |
398 |
|
Chr. 3 - 197003750 - 197004740 [+] |
View Full Product Details